ADVH49B83664 AChemBlock

1-(chloromethyl)-4-methoxy-2-(trifluoromethyl)benzene

  • Cas Number : 1260817-43-3
  • Molecular Weight : 224.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-chloromethyl-4-methoxy-2-trifluoromethyl-benzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89272
Availability details for
Quantity
  • Availability
  • Add to Cart