ADVH49B83665 AChemBlock

methyl 5-bromo-2-(dibromomethyl)benzoate

  • Cas Number : 1265286-99-4
  • Molecular Weight : 386.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-2-dibromomethyl-benzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89273
Availability details for
Quantity
  • Availability
  • Add to Cart