ADVH49B83667 AChemBlock

4-((tert-butoxycarbonyl)amino)-1-methyl-1H-imidazole-2-carboxylic acid

  • Cas Number : 128293-64-1
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-tert-butoxycarbonyl-amino-1-methyl-1h-imidazole-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89275
Availability details for
Quantity
  • Availability
  • Add to Cart