ADVH49B8366B AChemBlock

N-(2-(chloromethyl)phenyl)-2-methoxybenzamide

  • Molecular Weight : 275.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-2-chloromethyl-phenyl-2-methoxybenzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88659
Availability details for
Quantity
  • Availability
  • Add to Cart