ADVH49B8366C AChemBlock

4-nitrothiophene-2-carboxylic acid

  • Cas Number : 13138-70-0
  • Molecular Weight : 173.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-nitrothiophene-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89280
Availability details for
Quantity
  • Availability
  • Add to Cart