ADVH49B8366D AChemBlock

phenyl 4-amino-2-hydroxybenzoate

  • Cas Number : 133-11-9
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/phenyl-4-amino-2-hydroxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89281
Availability details for
Quantity
  • Availability
  • Add to Cart