ADVH49B8366E AChemBlock

2-(tert-butyl)-4-chlorophenol

  • Cas Number : 13395-85-2
  • Molecular Weight : 184.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-tert-butyl-4-chlorophenol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89282
Availability details for
Quantity
  • Availability
  • Add to Cart