ADVH49B83671 AChemBlock

2,4-bis(methoxymethoxy)-1-methylbenzene

  • Cas Number : 1380698-48-5
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-4-bis-methoxymethoxy-1-methylbenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89285
Availability details for
Quantity
  • Availability
  • Add to Cart