ADVH49B8367C AChemBlock

sodium hydrogen diphosphate

  • Cas Number : 14691-80-6
  • Molecular Weight : 243.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/sodium-hydrogen-diphosphate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89296
Availability details for
Quantity
  • Availability
  • Add to Cart