ADVH49B8367E AChemBlock

methyl 4-cyclopropylbenzoate

  • Cas Number : 148438-03-3
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-cyclopropylbenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89298
Availability details for
Quantity
  • Availability
  • Add to Cart