ADVH49B83683 AChemBlock

2-(phenylthio)benzoic acid

  • Cas Number : 1527-12-4
  • Molecular Weight : 230.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-phenylthio-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89303
Availability details for
Quantity
  • Availability
  • Add to Cart