ADVH49B83684 AChemBlock

4-(((2-ethylhexyl)oxy)carbonyl)benzoic acid

  • Cas Number : 155603-50-2
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-2-ethylhexyl-oxy-carbonyl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89304
Availability details for
Quantity
  • Availability
  • Add to Cart