ADVH49B83685 AChemBlock

methyl 1-methyl-5-nitro-1H-imidazole-2-carboxylate

  • Cas Number : 1563-98-0
  • Molecular Weight : 185.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-methyl-5-nitro-1h-imidazole-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89305
Availability details for
Quantity
  • Availability
  • Add to Cart