ADVH49B83686 AChemBlock

methyl 2-chloro-4-iodobenzoate

  • Cas Number : 156573-32-9
  • Molecular Weight : 296.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-chloro-4-iodobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89306
Availability details for
Quantity
  • Availability
  • Add to Cart