ADVH49B83688 AChemBlock

N,2-dihydroxy-4-methylbenzamide

  • Cas Number : 158671-29-5
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-2-dihydroxy-4-methylbenzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89308
Availability details for
Quantity
  • Availability
  • Add to Cart