ADVH49B8368A AChemBlock

5,6-dichloro-N-methoxy-N-methylnicotinamide

  • Cas Number : 162327-73-3
  • Molecular Weight : 235.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-6-dichloro-n-methoxy-n-methylnicotinamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89310
Availability details for
Quantity
  • Availability
  • Add to Cart