ADVH49B8368D AChemBlock

2,2'-(oxybis(ethane-2,1-diylsulfinyl))bis(ethan-1-ol)

  • Cas Number : 167940-02-5
  • Molecular Weight : 258.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-oxybis-ethane-2-1-diylsulfinyl-bis-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89313
Availability details for
Quantity
  • Availability
  • Add to Cart