ADVH49B8368E AChemBlock

5-bromo-2-(chloromethyl)pyridine

  • Cas Number : 168823-76-5
  • Molecular Weight : 206.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-chloromethyl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89314
Availability details for
Quantity
  • Availability
  • Add to Cart