ADVH49B8368F AChemBlock

bis(4-aminophenyl) terephthalate

  • Cas Number : 16926-73-1
  • Molecular Weight : 348.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/bis-4-aminophenyl-terephthalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89315
Availability details for
Quantity
  • Availability
  • Add to Cart