ADVH49B83693 AChemBlock

methyl 3-(2,4-dihydroxyphenyl)propanoate

  • Cas Number : 17422-90-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-2-4-dihydroxyphenyl-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89319
Availability details for
Quantity
  • Availability
  • Add to Cart