ADVH49B83694 AChemBlock

sodium 2-(2-aminophenyl)-2-oxoacetate

  • Cas Number : 17617-34-4
  • Molecular Weight : 187.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/sodium-2-2-aminophenyl-2-oxoacetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89320
Availability details for
Quantity
  • Availability
  • Add to Cart