ADVH49B83696 AChemBlock

ethyl 4-(benzyloxy)-3-hydroxybenzoate

  • Cas Number : 177429-27-5
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-benzyloxy-3-hydroxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89322
Availability details for
Quantity
  • Availability
  • Add to Cart