ADVH49B8369B AChemBlock

methyl 2-amino-5-hydroxybenzoate

  • Cas Number : 1882-72-0
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-amino-5-hydroxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89327
Availability details for
Quantity
  • Availability
  • Add to Cart