ADVH49B8369C AChemBlock

2-(6-((cyclopropylmethyl)carbamoyl)-2-(methoxycarbonyl)pyridin-3-yl)-4-methoxy-5-vinylbenzoic acid

  • Cas Number : 1883265-29-9
  • Molecular Weight : 410.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-6-cyclopropylmethyl-carbamoyl-2-methoxycarbonyl-pyridin-3-yl-4-methoxy-5-vinylbenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89328
Availability details for
Quantity
  • Availability
  • Add to Cart