ADVH49B8369D AChemBlock

ethyl 3-(benzyloxy)-4-hydroxybenzoate

  • Cas Number : 188564-63-8
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-benzyloxy-4-hydroxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89329
Availability details for
Quantity
  • Availability
  • Add to Cart