ADVH49B8369F AChemBlock

1-(methoxymethyl)-1H-imidazole

  • Cas Number : 20075-26-7
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methoxymethyl-1h-imidazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89331
Availability details for
Quantity
  • Availability
  • Add to Cart