ADVH49B836A1 AChemBlock

1-iodonaphthalen-2-ol

  • Cas Number : 2033-42-3
  • Molecular Weight : 270.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-iodonaphthalen-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89333
Availability details for
Quantity
  • Availability
  • Add to Cart