ADVH49B836A2 AChemBlock

(2R,3S,4R,5R)-2-(hydroxymethyl)-5-(6-((tetrahydrofuran-3-yl)amino)-9H-purin-9-yl)tetrahydrofuran-3,4-diol

  • Cas Number : 204512-90-3
  • Molecular Weight : 337.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-3s-4r-5r-2-hydroxymethyl-5-6-tetrahydrofuran-3-yl-amino-9h-purin-9-yl-tetrahydrofuran-3-4-diol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89334
Availability details for
Quantity
  • Availability
  • Add to Cart