ADVH49B836A3 AChemBlock

(9H-fluoren-9-yl)methyl (4-aminophenyl)carbamate

  • Cas Number : 205688-13-7
  • Molecular Weight : 330.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/9h-fluoren-9-yl-methyl-4-aminophenyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89335
Availability details for
Quantity
  • Availability
  • Add to Cart