ADVH49B836A5 AChemBlock

4-aminophenyl 4-aminobenzoate

  • Cas Number : 20610-77-9
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-aminophenyl-4-aminobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89337
Availability details for
Quantity
  • Availability
  • Add to Cart