ADVH49B836A9 AChemBlock

O-(2-fluorobenzyl)hydroxylammonium chloride

  • Cas Number : 215599-91-0
  • Molecular Weight : 177.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/o-2-fluorobenzyl-hydroxylammonium-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89341
Availability details for
Quantity
  • Availability
  • Add to Cart