ADVH49B836AB AChemBlock

methyl 1-(2,6-difluorobenzyl)-1H-1,2,3-triazole-4-carboxylate

  • Cas Number : 217448-86-7
  • Molecular Weight : 253.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-2-6-difluorobenzyl-1h-1-2-3-triazole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89343
Availability details for
Quantity
  • Availability
  • Add to Cart