ADVH49B836B2 AChemBlock

1-benzyl-5-nitro-1H-indazole

  • Cas Number : 23856-20-4
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-5-nitro-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89350
Availability details for
Quantity
  • Availability
  • Add to Cart