ADVH49B836BF AChemBlock

1-benzyl-3-bromobenzene

  • Cas Number : 27798-39-6
  • Molecular Weight : 247.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-3-bromobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89363
Availability details for
Quantity
  • Availability
  • Add to Cart