ADVH49B836C2 AChemBlock

methyl 2-(5-nitropyridin-2-yl)acetate

  • Cas Number : 292600-22-7
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-5-nitropyridin-2-yl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89366
Availability details for
Quantity
  • Availability
  • Add to Cart