ADVH49B836C6 AChemBlock

2-(benzylsulfonyl)-1-(1H-indol-3-yl)-1,2-dihydroisoquinoline

  • Cas Number : 313526-24-8
  • Molecular Weight : 400.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-benzylsulfonyl-1-1h-indol-3-yl-1-2-dihydroisoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89370
Availability details for
Quantity
  • Availability
  • Add to Cart