ADVH49B836C7 AChemBlock

5-(3,4-dimethoxybenzyl)-1,3,4-thiadiazol-2-amine

  • Cas Number : 313957-85-6
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-3-4-dimethoxybenzyl-1-3-4-thiadiazol-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89371
Availability details for
Quantity
  • Availability
  • Add to Cart