ADVH49B836CC AChemBlock

2-(benzo[d][1,3]dioxol-5-yl)acetate

  • Cas Number : 326-58-9
  • Molecular Weight : 179.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-benzo-d-1-3-dioxol-5-yl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89377
Availability details for
Quantity
  • Availability
  • Add to Cart