ADVH49B836CD AChemBlock

N1,N1,N2-trimethylethane-1,2-diamine dihydrochloride

  • Cas Number : 326888-32-8
  • Molecular Weight : 175.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n1-n1-n2-trimethylethane-1-2-diamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89378
Availability details for
Quantity
  • Availability
  • Add to Cart