ADVH49B836D0 AChemBlock

methyl 3-hydroxy-4,5-dimethylthiophene-2-carboxylate

  • Cas Number : 32822-84-7
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-hydroxy-4-5-dimethylthiophene-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89381
Availability details for
Quantity
  • Availability
  • Add to Cart