ADVH49B836D1 AChemBlock

4,4-diethoxy-1,1,1-trifluorobutan-2-one

  • Cas Number : 333339-64-3
  • Molecular Weight : 214.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-4-diethoxy-1-1-1-trifluorobutan-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89382
Availability details for
Quantity
  • Availability
  • Add to Cart