ADVH49B836D2 AChemBlock

3-(2-methoxyphenyl)propanal

  • Cas Number : 33538-83-9
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-2-methoxyphenyl-propanal.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89383
Availability details for
Quantity
  • Availability
  • Add to Cart