ADVH49B836D5 AChemBlock

2-methyl-3-nitroimidazo[1,2-a]pyridine

  • Cas Number : 34165-09-8
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-3-nitroimidazo-1-2-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89386
Availability details for
Quantity
  • Availability
  • Add to Cart