ADVH49B836D6 AChemBlock

1-ethoxy-3-methyl-2-nitrobenzene

  • Cas Number : 342044-61-5
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-ethoxy-3-methyl-2-nitrobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89387
Availability details for
Quantity
  • Availability
  • Add to Cart