ADVH49B836D7 AChemBlock

6-nitroindoline-2,3-dione

  • Cas Number : 3433-54-3
  • Molecular Weight : 192.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-nitroindoline-2-3-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89388
Availability details for
Quantity
  • Availability
  • Add to Cart