ADVH49B836DA AChemBlock

sodium (4-nitrophenyl)methanesulfonate

  • Cas Number : 36639-50-6
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/sodium-4-nitrophenyl-methanesulfonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89391
Availability details for
Quantity
  • Availability
  • Add to Cart