ADVH49B836DC AChemBlock

2-benzyl-2,3-dihydroisoquinolin-4(1H)-one hydrochloride

  • Cas Number : 37481-69-9
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-benzyl-2-3-dihydroisoquinolin-4-1h-one-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89393
Availability details for
Quantity
  • Availability
  • Add to Cart