ADVH49B836DE AChemBlock

4,5,6,7-tetrahydro-1H-benzo[d]imidazole

  • Cas Number : 3752-24-7
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-5-6-7-tetrahydro-1h-benzo-d-imidazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89395
Availability details for
Quantity
  • Availability
  • Add to Cart