ADVH49B836E6 AChemBlock

1-(4-(benzyloxy)phenyl)-2-bromoethan-1-one

  • Cas Number : 4254-67-5
  • Molecular Weight : 305.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-benzyloxy-phenyl-2-bromoethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89405
Availability details for
Quantity
  • Availability
  • Add to Cart