ADVH49B836E8 AChemBlock

N'-isopropylacetohydrazide

  • Cas Number : 4466-50-6
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-isopropylacetohydrazide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89407
Availability details for
Quantity
  • Availability
  • Add to Cart